missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Expi293™ PRO Expression Medium

Numéro de catalogue. a4000135504
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A4000135504 20 L
A4000135501 1 L
A4000135505 6 x 1 L
A4000135503 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. A4000135504 Fournisseur Gibco™ Code fournisseur A4000135504
Il en reste null

Expi293 PRO Expression Medium supports rapid, high-yield protein production from HEK293 cells. Designed for scalable, high-density cultures with consistent expression across a variety of proteins, including complex and low-yield targets.

Gibco Expi293 PRO Expression Medium is a chemically defined, serum-free, protein-free medium for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells. It is specifically designed as a core component to support high density culture of Expi293 PRO cells in the Expi293 PRO Expression System for scalable transient protein expression.

  • Supports growth of suspension HEK 293 cultures to densities of over 15 x 10^6 cells/mL
  • Transfection-compatible serum-free suspension culture medium
  • Helps enable sustained, high level expression of high density transiently transfected cultures, achieving yields of up to 5 g/L of recombinant protein
  • Supports growth and transfection of HEK293 cells in culture formats of 1 mL to 10 L
  • Does not contain phenol red

Expi293 PRO Expression Medium is formulated with GlutaMAX-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. Expi293 PRO Expression Medium contains no human or animal-origin components. The medium is not recommended for adherent cell cultures.

Spécifications

Cell Line 293 (HEK)
Cell Type Embryonic Kidney Cells
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Liquid
Product Type Expression Medium
Serum Level Serum-free
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives GlutaMAX
Without Additives No HEPES, No Phenol Red
Product Line Expi293 PRO
Quantity 20 L
Content And Storage Store in the dark at +2°C to +8°C
Culture Type Suspension Cell Culture
Afficher plus de résultats Afficher moins de résultats

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.