Learn More
New England Biolab Ltd Exonuclease VIII, truncated 1000 units
Numéro de catalogue. 501526396
Encompass
Exonuclease VIII, truncated, is able to initiate nucleotide removal from the 5' termini of linear double-stranded DNA in the 5' to 3' direction.
Numéro de catalogue. 501526396 Fournisseur New England Biolab Ltd Code fournisseur M0545S
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Exonuclease VIII, truncated, is able to initiate nucleotide removal from the 5' termini of linear double-stranded DNA in the 5' to 3' direction.