Learn More
New England Biolab Ltd Exonuclease T 1250 units
Numéro de catalogue. 50811692
Encompass
Exonuclease T (Exo T), also known as RNase T, is a single-stranded RNA or DNA specific nuclease that requires a free 3' terminus and removes nucleotides in the 3' to 5' direction. Exonuclease T can be used to generate blunt ends from RNA or DNA molecules that have 3 extensions.
Numéro de catalogue. 50811692 Fournisseur New England Biolab Ltd Code fournisseur M0265L
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Exonuclease T (Exo T), also known as RNase T, is a single-stranded RNA or DNA specific nuclease that requires a free 3' terminus and removes nucleotides in the 3' to 5' direction. Exonuclease T can be used to generate blunt ends from RNA or DNA molecules that have 3 extensions.