missing translation for 'onlineSavingsMsg'
Learn More

Excelta™ Micro Tip Probes

With smallest available tips for SMT and other micro assembly applications

Supplier:  Excelta™ 332C

 Afficher plus de versions de ce produit

Numéro de catalogue. 17467611


Il en reste null

Description

Description

  • Stainless-steel micro tips for high-precision work
  • Length: 6.5 (16.5cm)
  • Tip size: 1mil
  • Points are hardened for long life
Spécifications

Identifiants chimiques

Micro Tip Probe
For SMT and other micro assembly applications
6.5 in.
Stainless Steel
1 mm
16.5 cm

Spécifications

Micro Tip Probe
For SMT and other micro assembly applications
1 mm
16.5 cm
Stainless Steel
25° angled
6.5 in.
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions