missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EWS Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
EWSR1 is a multifunctional protein that is involved in various cellular processes, including gene expression, cell signaling, and RNA processing and transport. The protein includes an N-terminal transcriptional activation domain and a C-terminal RNA-binding domain.
Spécifications
Spécifications
| Antigène | EWS |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugué | Unconjugated |
| Formule | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gène | EWSR1 |
| Numéro d’acquisition du gène | Q01844 |
| Alias du gène | AC002059.7, EWS, bK984G1.4 |
| Symboles de gène | EWSR1 |
| Show More |