missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) phosphate hydrate, 99.9% (REO)

Catalog No. p-7051790
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4198903 1 g
AA4198906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4198903 Supplier Thermo Scientific Chemicals Supplier No. 04198903

CAS: 14913-20-3 | EuO4P | 246.93 g/mol

Europium finds application in optical coating and thin film application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14913-20-3
Molecular Formula EuO4P
Molecular Weight (g/mol) 246.93
MDL Number MFCD05664261
InChI Key KJNZTHUWRRLWOA-UHFFFAOYSA-K
Synonym europium iii phosphate hydrate, europium 3+ hydrate phosphate, europium 3+ phosphate-water 1/1/1, europium 3+ ion hydrate phosphate
PubChem CID 16217374
SMILES [Eu+3].[O-]P([O-])([O-])=O

Specifications

Physical Form Chunks and/or Powder
Quantity 1 g
Linear Formula EuPO4·xH2O
Solubility Information Insoluble in water.
IUPAC Name europium(3+) phosphate
Formula Weight 246.93 (Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium(III) phosphate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-901-2
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only