missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) Chloride Hexahydrate 96.0+%, TCI America™

Catalog No. E07075G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E07075G Supplier TCI America Supplier No. E07075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13759-92-7
Molecular Formula Cl3Eu
MDL Number MFCD00149701

Specifications

Color White
Physical Form Crystalline Powder
Quantity 5 g
Formula Weight 258.31
Percent Purity ≥96.0% (T)
Chemical Name or Material Europium(III) Chloride Hexahydrate
RTECSNumber LE7600000
TSCA No