missing translation for 'onlineSavingsMsg'
Learn More

Europium ICP Standard, 1000 ppm Eu in 3% HNO3, Ricca Chemical

Catalog No. PEU1KN100
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
PEU1KN100 Natural LDPE Bottle
PEU1KN500 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PEU1KN100 Supplier Ricca Chemical Company Supplier No. PEU1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1308-96-9 , 7732-18-5 , 1308-96-9
Molecular Formula Eu2O3
Molecular Weight (g/mol) 351.925
InChI Key AEBZCFFCDTZXHP-UHFFFAOYSA-N
PubChem CID 159371
IUPAC Name europium(3+);oxygen(2-)
SMILES [O-2].[O-2].[O-2].[Eu+3].[Eu+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Eu
CAS Min % 93.0, 4.56, 0.11
CAS Max % 97.0, 4.74, 0.11
Equivalence 1 mL = 1 mg Eu
Chemical Name or Material Europium ICP Standard