missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eugenol Acetate 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Boiling Point | 164°C |
| Quantity | 25 g |
| Formula Weight | 206.24 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | Eugenol Acetate |
Safety and Handling
| EINECSNumber | (3)-0640 |
| RTECSNumber | SJ4550000 |
| TSCA | Yes |