missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyltriphenylphosphonium Bromide 98.0+%, TCI America™
Supplier: TCI America E0382100G
Specifications
| Ethyltriphenylphosphonium Bromide | |
| 210°C | |
| C20H20BrP | |
| 100 g | |
| ethyltriphenylphosphonium bromide, ethyl triphenyl phosphonium bromide, triphenylethylphosphonium bromide, phosphonium, ethyltriphenyl-, bromide, ethyltriphenylphosphanium bromide, ethyl triphenylphosphonium bromide, unii-r85v84ul5v, ethyltriphenylphosphoniumbromide, guaifenesin powder, pubchem18705 | |
| [Br-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 371.26 | |
| 371.26 | |
| Crystalline Powder |
| 1530-32-1 | |
| White-Yellow | |
| MFCD00011838 | |
| 2811 | |
| JHYNXXDQQHTCHJ-UHFFFAOYSA-M | |
| ethyltriphenylphosphanium bromide | |
| 73727 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 1530-32-1 | |
| 371.26 | |
| JHYNXXDQQHTCHJ-UHFFFAOYSA-M | |
| 73727 | |
| [Br-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C20H20BrP | |
| MFCD00011838 | |
| ethyltriphenylphosphonium bromide, ethyl triphenyl phosphonium bromide, triphenylethylphosphonium bromide, phosphonium, ethyltriphenyl-, bromide, ethyltriphenylphosphanium bromide, ethyl triphenylphosphonium bromide, unii-r85v84ul5v, ethyltriphenylphosphoniumbromide, guaifenesin powder, pubchem18705 | |
| ethyltriphenylphosphanium bromide |
Safety and Handling
TSCA : Yes