missing translation for 'onlineSavingsMsg'
Learn More

Ethyltriphenylphosphonium Bromide 98.0+%, TCI America™

Catalog No. p-7128972
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
E038225G 25 g
E0382100G 100 g
E0382500G 500 g
3 options

Catalog No. E038225G

Supplier: TCI America E038225G

Only null left

Chemical Identifiers

CAS 1530-32-1
Molecular Formula C20H20BrP
Molecular Weight (g/mol) 371.26
MDL Number MFCD00011838
InChI Key JHYNXXDQQHTCHJ-UHFFFAOYSA-M
Synonym ethyltriphenylphosphonium bromide, ethyl triphenyl phosphonium bromide, triphenylethylphosphonium bromide, phosphonium, ethyltriphenyl-, bromide, ethyltriphenylphosphanium bromide, ethyl triphenylphosphonium bromide, unii-r85v84ul5v, ethyltriphenylphosphoniumbromide, guaifenesin powder, pubchem18705
PubChem CID 73727
IUPAC Name ethyltriphenylphosphanium bromide
SMILES [Br-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 210°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 371.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyltriphenylphosphonium Bromide