missing translation for 'onlineSavingsMsg'
Learn More

Ethylparaben Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012747
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012747 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012747 Supplier Merck Emd Millipore Supplier No. PHR10111G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 120-47-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula HOC6H4 CO2C2H5
MDL Number MFCD00002353
Melting Point 114°C to 117°C
Boiling Point 297°C to 298°C
Synonym Ethyl 4-hydroxybenzoate; Ethylparaben
Type Certified Reference Material
Quantity 1 g
Formula Weight 166.17
Grade Certified Reference Material
Show More Show Less