missing translation for 'onlineSavingsMsg'
Learn More

Ethylparaben, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111012748
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012748 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012748 Supplier Merck Emd Millipore Supplier No. SGMAPHR101110G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 120-47-8
Physical Form Neat
Linear Formula HOC6H4CO2C2H5
MDL Number MFCD00002353
Melting Point 114°C to 117°C
Boiling Point 297°C to 298°C
Synonym Ethyl 4-hydroxybenzoate; Ethylparaben
Quantity 10 g
Formula Weight 166.17
Grade Certified Reference Material