missing translation for 'onlineSavingsMsg'
Learn More

Ethylmagnesium chloride, 2.7M (25 wt.%) solution in THF, AcroSeal™

Catalog No. AC252571000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC252571000 100 mL AcroSeal™ Glass bottle
AC252578000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC252571000 Supplier Thermo Scientific Chemicals Supplier No. 252571000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2386-64-3 | C2H5ClMg | 88.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2386-64-3 , 109-99-9
Molecular Formula C2H5ClMg
Molecular Weight (g/mol) 88.82
MDL Number MFCD00000472
InChI Key ZWNJJACKPNTXHY-UHFFFAOYSA-M
Synonym ethylmagnesium chloride, chloroethylmagnesium, ethylmagnesiumchloride, magnesium, chloroethyl, etmgcl, chloro ethyl magnesium, ethyl magnesium chloride, chloroethyl-magnesiu, ethyl magnesiumchloride, ethylmagnesium chloride solution
PubChem CID 75432
IUPAC Name magnesium;ethane;chloride
SMILES CC[Mg]Cl

Specifications

Color Brown
Density 1.0000g/mL
Packaging AcroSeal™ Glass bottle
Linear Formula C2H5MgCl
Quantity 100 mL
Specific Gravity 1
Solubility Information Solubility in water: reacts
Formula Weight 88.83
Percent Purity 24 to 26%
Physical Form Liquid
Chemical Name or Material Ethylmagnesium chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flammable liquid and vapor.
Reacts violently w
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture
EINECSNumber 219-206-9
TSCA TSCA

RUO – Research Use Only