missing translation for 'onlineSavingsMsg'
Learn More

Ethylhexadecyldimethylammonium Bromide, High Purity, 99%, Spectrum™ Chemical

$259.96 - $35764.71

Chemical Identifiers

CAS 124-03-8
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610454
View Documents
Spectrum Chemical Mfg Cor
E10545GM
5 g Amber Glass Bottle
Each for $259.96
Only null left
 
18610455
View Documents
Spectrum Chemical Mfg Cor
E1054100GM
100 g Poly Bottle
Each for $569.71
Only null left
 
18610456
View Documents
Spectrum Chemical Mfg Cor
E1054500GM
500 g Poly Bottle
Each for $2,000.21
Only null left
 
18610457
View Documents
Spectrum Chemical Mfg Cor
E105425KG
2.5 kg Poly Pail
Each for $6,217.71
Only null left
 
18610458
View Documents
Spectrum Chemical Mfg Cor
E105412KGBL
12 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18610459
View Documents
Spectrum Chemical Mfg Cor
E105425KGBL
25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

124-03-8

Specifications

124-03-8
Amber Glass Bottle
0.99
1
5 g
High Purity
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.