missing translation for 'onlineSavingsMsg'
Learn More

Ethylhexadecyldimethylammonium Bromide, High Purity, 99%, Spectrum™ Chemical

Catalog No. 18610454
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610454 5 g Amber Glass Bottle
18610455 100 g Amber Glass Bottle
18610456 500 g Amber Glass Bottle
18610457 2.5 kg Amber Glass Bottle
18610458 12 kg Amber Glass Bottle
18610459 25 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610454 Supplier Spectrum Chemical Mfg Cor Supplier No. E10545GM

Chemical Identifiers

CAS 124-03-8

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 0.99
Grade High Purity
Physical Form Powder or Waxy Solid
Chemical Name or Material Ethylhexadecyldimethylammonium Bromide

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.