missing translation for 'onlineSavingsMsg'
Learn More

Ethylhexadecyldimethylammonium Bromide, High Purity, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor E1054500GM

 View more versions of this product

Catalog No. 18610456


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

124-03-8
Poly Bottle
0.99
1
500 g
High Purity

Chemical Identifiers

124-03-8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.