missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylhexadecyldimethylammonium Bromide, High Purity, 99%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 12 kg |
| Percent Purity | 0.99 |
| Grade | High Purity |
| Physical Form | Powder or Waxy Solid |
| Chemical Name or Material | Ethylhexadecyldimethylammonium Bromide |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.