missing translation for 'onlineSavingsMsg'
Learn More

Ethylferrocene, 98%, Thermo Scientific Chemicals

Catalog No. AA3925309
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3925309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3925309 Supplier Thermo Scientific Chemicals Supplier No. 03925309

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 1273-89-8
Density 1.256 g/mL
Boiling Point 264°C
Molecular Formula C12H14Fe
Refractive Index 1.601
Quantity 10 g
MDL Number MFCD00059071
InChI Key FCNXGBYXGSKCDG-UHFFFAOYSA-N
SMILES [Fe++].[CH-]1C=CC=C1.CC[C-]1C=CC=C1
IUPAC Name λ²-iron(2+) 1-ethylcyclopenta-2,4-dien-1-ide cyclopenta-2,4-dien-1-ide
Molecular Weight (g/mol) 214.09
Formula Weight 214.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethylferrocene
Show More Show Less
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 215-056-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only