missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Trisodium Salt Hydrate, MP Biomedicals™

Catalog No. ICN15108891
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15108891 1 kg
ICN15108890 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15108891 Supplier MP Biomedicals Inc Supplier No. 0215108891

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Water soluble. Clear colorless to light yellow solution at 32gm plus 100mL of 1M sodium hydroxide

Chemical Identifiers

CAS 85715-60-2
Molecular Formula C10H18N2Na3O9
Molecular Weight (g/mol) 379.228
MDL Number MFCD00149675
InChI Key DFCJTQJARZUURB-UHFFFAOYSA-N
Synonym edta, trisodium salt trihydrate
PubChem CID 131865091
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;hydrate
SMILES C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.[Na].[Na].[Na]

Specifications

Color White
Quantity 1 kg
Formula Weight 358.19
Percent Purity ≥95%
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic Acid Trisodium Salt Hydrate
EINECSNumber 200-449-4