missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, Electrophoresis Grade, 99.4+%

Catalog No. AAJ6558522
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6558522 100 g
AAJ6558536 500 g
AAJ65585A4 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6558522 Supplier Thermo Scientific Chemicals Supplier No. J6558522

CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Melting Point ∽245°C (decomposition)
Color White
Density 0.68
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Quantity 100 g
Beilstein 1716295
Merck Index 143517
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
Formula Weight 292.24
Percent Purity ≥99.4%
Grade Electrophoresis
Physical Form Powder
Chemical Name or Material Ethylenediaminetetraacetic acid
Show More Show Less
Hazard Category Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after repeated exposure (category 2)
Hazard Statement Causes serious eye irritation, Harmful if inhaled, May cause damage to organs through prolonged or repeated exposure if inhaled
EINECSNumber 200-449-4
RTECSNumber AH4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only