missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid disodium magnesium salt hydrate

Catalog No. AAL1185722
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1185722 100 g
AAL1185714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1185722 Supplier Thermo Scientific Chemicals Supplier No. L1185722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 194491-32-2 | C10H12MgN2Na2O8 | 358.50 g/mol

Ethylenediaminetetraacetic acid disodium magnesium salt hydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is used as chelating agents over a broad pH range in aqueous systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is used as chelating agents over a broad pH range in aqueous systems.

Solubility
Soluble in water. Insoluble in acid and organic liquids

Notes
Incompatible with oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Store at room temperature.

Chemical Identifiers

CAS 194491-32-2
Molecular Formula C10H12MgN2Na2O8
Molecular Weight (g/mol) 358.50
MDL Number MFCD00078217
InChI Key AWNVVAMWLMUZOZ-UHFFFAOYSA-J
Synonym unii-ndt563s5vz, edta magnesium disodium, magnesium disodium edta, ndt563s5vz, magnesium disodium ethylenediaminetetraacetate, edta magnesium disodium salt hydrate, ethylenediaminetetraacetic acid magnesium disodium salt, ethylenediaminetetraacetic acid disodium magnesium salt, ethylenediaminetetraacetic acid disodium magnesium salt hydrate, magnesium sodium ethylenediaminetetraacetate
PubChem CID 161064
IUPAC Name magnesium;disodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate
SMILES [Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 100 g
Solubility Information Soluble in water. Insoluble in acid and organic liquids
Formula Weight 358.50 (Anhydrous)
Chemical Name or Material Ethylenediaminetetraacetic acid disodium magnesium salt hydrate
EINECSNumber 238-372-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only