missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0), Fisher BioReagents

Catalog No. BP2482100
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
20 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Poly CUBE
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2482100 100 mL Poly Bottle
BP2482500 500 mL Poly Bottle
BP24821 1 L Nalgene Poly Bottle
BP248220 20 L Poly CUBE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BP2482100 Supplier Fisher BioReagents Supplier No. BP2482100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

EDTA is used extensively in molecular biology to minimize metal ion impurities in reaction buffers. Ideal for DNA work.

Chemical Identifiers

CAS 60-00-4 , 7732-18-5
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Color Colorless
Linear Formula (HOOCCH2)2NCH2CH2N(CH2COOH)2
Concentration 0.475 to 0.525M
Merck Index 15, 3565
Formula Weight 292.25
Identification Pass Test
Packaging Poly Bottle
pH 8.0
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Ethylenediamine Tetraacetic Acid, 0.5 M Solution, pH 8.0 (for DNA Work)
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT