missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Éthylène glycol 99,5+%, TCI America™
Spécifications
Spécifications
| Physical Form | Liquide clair à 20 °C |
| Melting Point | -13°C |
| Boiling Point | 196°C |
| Color | Incolore |
| Quantity | 25 g |
| Formula Weight | 62.07 |
| Percent Purity | ≥99.5% (GC) |
| Chemical Name or Material | Ethylene Glycol |
Sécurité et manipulation
| EINECSNumber | (2)-0230 |
| RTECSNumber | KW2975000 |
| TSCA | Yes |