missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol Solution, NMR Reference Standard, 80% in DMSO-d6 (99.9 atom% D), NMR tube size 3 mm 8 in., MilliporeSigma™ Supelco™

Catalog No. 111012408
Change view
Click to view available options
Quantity:
0.235 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012408 0.235 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012408 Supplier Merck Emd Millipore Supplier No. 7219808IN

Specifications

Packaging 3 mm O.D. tube
CAS 107-21-1
Linear Formula C2H6O2
MDL Number MFCD00002885
Type NMR Reference Standard
Quantity 0.235 mL
Formula Weight 62.07
Grade NMR Reference Standard
Concentration or Composition (by Analyte or Components) 80% in DMSO- d6 (99.9 atom % D)