missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Monopropyl Ether 98.0+%, TCI America™

Catalog No. E0514500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0514500ML 500 mL
E051425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0514500ML Supplier TCI America Supplier No. E0514500ML

Chemical Identifiers

CAS 2807-30-9
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.149
MDL Number MFCD00072645
InChI Key YEYKMVJDLWJFOA-UHFFFAOYSA-N
Synonym ethanol, 2-propoxy, ethylene glycol monopropyl ether, propyl cellosolve, propoxyethanol, propyl glycol, 2-propoxyethan-1-ol, ektasolve ep, 2-propyloxy ethanol, n-propoxyethanol, ethylene glycol mono-n-propyl ether
PubChem CID 17756
IUPAC Name 2-propoxyethanol
SMILES CCCOCCO

Specifications

Color Colorless
Boiling Point 151°C
Quantity 500 mL
UN Number 1992
Formula Weight 104.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Monopropyl Ether
EINECSNumber (2)-2424&(7)-0097
RTECSNumber KM2800000
TSCA Yes