missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Mono-p-tolyl Ether 98.0+%, TCI America™
$71.23 - $229.10
Chemical Identifiers
| CAS | 15149-10-7 |
|---|---|
| Molecular Formula | C9H12O2 |
| Molecular Weight (g/mol) | 152.193 |
| MDL Number | MFCD00020600 |
| InChI Key | FFWXHQFJNOGDJE-UHFFFAOYSA-N |
| Synonym | 4-(2-Hydroxyethoxy)toluene, 2-(4-Methylphenoxy)ethanol |
| PubChem CID | 84804 |
| IUPAC Name | 2-(4-methylphenoxy)ethanol |
| SMILES | CC1=CC=C(C=C1)OCCO |
Chemical Identifiers
| 15149-10-7 | |
| 152.193 | |
| FFWXHQFJNOGDJE-UHFFFAOYSA-N | |
| 84804 | |
| CC1=CC=C(C=C1)OCCO |
| C9H12O2 | |
| MFCD00020600 | |
| 4-(2-Hydroxyethoxy)toluene, 2-(4-Methylphenoxy)ethanol | |
| 2-(4-methylphenoxy)ethanol |
Specifications
| 15149-10-7 | |
| White-Yellow | |
| C9H12O2 | |
| 5 g | |
| FFWXHQFJNOGDJE-UHFFFAOYSA-N | |
| 2-(4-methylphenoxy)ethanol | |
| 84804 | |
| ≥98.0% (GC) | |
| Ethylene Glycol Mono-p-tolyl Ether |
| 45°C | |
| 123°C | |
| MFCD00020600 | |
| 4-(2-Hydroxyethoxy)toluene, 2-(4-Methylphenoxy)ethanol | |
| CC1=CC=C(C=C1)OCCO | |
| 152.193 | |
| 152.19 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2777
TSCA : Yes