missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Mono-p-tolyl Ether 98.0+%, TCI America™

Supplier:  TCI America E03035G

 View more versions of this product

Catalog No. E03035G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethylene Glycol Mono-p-tolyl Ether
45°C
123°C
MFCD00020600
4-(2-Hydroxyethoxy)toluene, 2-(4-Methylphenoxy)ethanol
CC1=CC=C(C=C1)OCCO
152.193
152.19
Crystalline Powder
15149-10-7
White-Yellow
C9H12O2
5 g
FFWXHQFJNOGDJE-UHFFFAOYSA-N
2-(4-methylphenoxy)ethanol
84804
≥98.0% (GC)

Chemical Identifiers

15149-10-7
152.193
FFWXHQFJNOGDJE-UHFFFAOYSA-N
84804
CC1=CC=C(C=C1)OCCO
C9H12O2
MFCD00020600
4-(2-Hydroxyethoxy)toluene, 2-(4-Methylphenoxy)ethanol
2-(4-methylphenoxy)ethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2777

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions