accessibility menu, dialog, popup

UserName

CAS RN 15149-10-7

CH3OOH

CAS RN 15149-10-7

IUPAC Name: 2-(4-methylphenoxy)ethan-1-ol
Synonyms: methylphenoxy-ethanol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: FFWXHQFJNOGDJE-UHFFFAOYSA-N
SMILES: CC1=CC=C(OCCO)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results