missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Diacetate 97.0+%, TCI America™

Catalog No. E0109500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0109500ML 500 mL
E010925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0109500ML Supplier TCI America Supplier No. E0109500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-55-7
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00008718
InChI Key JTXMVXSTHSMVQF-UHFFFAOYSA-N
Synonym ethylene glycol diacetate, 1,2-diacetoxyethane, glycol diacetate, ethylene diacetate, ethanediol diacetate, 1,2-ethanediol, diacetate, 1,2-ethanediol diacetate, ethylene glycol, diacetate, ethylene diethanoate, aptex donor h-plus
PubChem CID 8121
IUPAC Name 2-acetyloxyethyl acetate
SMILES CC(=O)OCCOC(=O)C

Specifications

Color Colorless
Boiling Point 190°C
Quantity 500 mL
Formula Weight 146.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Diacetate
EINECSNumber (2)-0665
RTECSNumber KW4025000
TSCA Yes