missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol Di(m-tolyl) Ether 97.0+%, TCI America™
$29.83 - $57.79
Chemical Identifiers
| CAS | 54914-85-1 |
|---|---|
| Molecular Formula | C16H18O2 |
| Molecular Weight (g/mol) | 242.32 |
| MDL Number | MFCD00025952 |
| InChI Key | OAGNKYSIOSDNIG-UHFFFAOYSA-N |
| Synonym | 1,1′-[1,2-Ethanediylbis(oxy)]bis(3-methylbenzene), 1,2-Bis(3-methylphenoxy)ethane |
| PubChem CID | 97354 |
| IUPAC Name | 1-methyl-3-[2-(3-methylphenoxy)ethoxy]benzene |
| SMILES | CC1=CC(OCCOC2=CC(C)=CC=C2)=CC=C1 |
Chemical Identifiers
| 54914-85-1 | |
| 242.32 | |
| OAGNKYSIOSDNIG-UHFFFAOYSA-N | |
| 97354 | |
| CC1=CC(OCCOC2=CC(C)=CC=C2)=CC=C1 |
| C16H18O2 | |
| MFCD00025952 | |
| 1,1′-[1,2-Ethanediylbis(oxy)]bis(3-methylbenzene), 1,2-Bis(3-methylphenoxy)ethane | |
| 1-methyl-3-[2-(3-methylphenoxy)ethoxy]benzene |
Specifications
| 54914-85-1 | |
| White-Yellow | |
| MFCD00025952 | |
| 3077 | |
| OAGNKYSIOSDNIG-UHFFFAOYSA-N | |
| 1-methyl-3-[2-(3-methylphenoxy)ethoxy]benzene | |
| 97354 | |
| ≥97.0% (GC) | |
| Ethylene Glycol Di(m-tolyl) Ether |
| 98°C | |
| C16H18O2 | |
| 5 g | |
| 1,1′-[1,2-Ethanediylbis(oxy)]bis(3-methylbenzene), 1,2-Bis(3-methylphenoxy)ethane | |
| CC1=CC(OCCOC2=CC(C)=CC=C2)=CC=C1 | |
| 242.32 | |
| 242.32 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-3886
TSCA : Yes