missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Di(m-tolyl) Ether 97.0+%, TCI America™

Supplier:  TCI America E133525G

 View more versions of this product

Catalog No. E133525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethylene Glycol Di(m-tolyl) Ether
98°C
C16H18O2
25 g
1,1′-[1,2-Ethanediylbis(oxy)]bis(3-methylbenzene), 1,2-Bis(3-methylphenoxy)ethane
CC1=CC(OCCOC2=CC(C)=CC=C2)=CC=C1
242.32
242.32
Crystalline Powder
54914-85-1
White-Yellow
MFCD00025952
3077
OAGNKYSIOSDNIG-UHFFFAOYSA-N
1-methyl-3-[2-(3-methylphenoxy)ethoxy]benzene
97354
≥97.0% (GC)

Chemical Identifiers

54914-85-1
242.32
OAGNKYSIOSDNIG-UHFFFAOYSA-N
97354
CC1=CC(OCCOC2=CC(C)=CC=C2)=CC=C1
C16H18O2
MFCD00025952
1,1′-[1,2-Ethanediylbis(oxy)]bis(3-methylbenzene), 1,2-Bis(3-methylphenoxy)ethane
1-methyl-3-[2-(3-methylphenoxy)ethoxy]benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3886

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions