CAS RN 54914-85-1
CAS RN 54914-85-1
IUPAC Name:
1-methyl-3-[2-(3-methylphenoxy)ethoxy]benzene
Synonyms:
1-methyl-3-[2-(3-methylphenoxy)ethoxy]benzene
Molecular Weight (g/mol):
242.32
Molecular Formula:
C16H18O2
InChi Key:
OAGNKYSIOSDNIG-UHFFFAOYSA-N
SMILES:
CC1=CC(OCCOC2=CC(C)=CC=C2)=CC=C1
1
–
1
of
1
results
1
Ethylene Glycol Di(m-tolyl) Ether 97.0+%, TCI America™
CAS: 54914-85-1 Molecular Formula: C16H18O2 Molecular Weight (g/mol): 242.32 MDL Number: MFCD00025952 InChI Key: OAGNKYSIOSDNIG-UHFFFAOYSA-N Synonym: 1,1′-[1,2-Ethanediylbis(oxy)]bis(3-methylbenzene), 1,2-Bis(3-methylphenoxy)ethane PubChem CID: 97354 IUPAC Name: 1-methyl-3-[2-(3-methylphenoxy)ethoxy]benzene SMILES: CC1=CC(OCCOC2=CC(C)=CC=C2)=CC=C1