missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol-bis-(β-aminoethylether)-N,N,N',N'-tetraacetic acid, 99.6%, MP Biomedicals™

Catalog No. ICN19517490
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19517490 500 g
ICN19517410 10 g
ICN19517425 25 g
ICN19517483 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19517490 Supplier MP Biomedicals Inc Supplier No. 0219517490

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 67-42-5 | C14H24N2O10 | 380.35 g/mol

Chemical Identifiers

CAS 67-42-5
Molecular Formula C14H24N2O10
Molecular Weight (g/mol) 380.35
InChI Key DEFVIWRASFVYLL-UHFFFAOYSA-N
Synonym egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
PubChem CID 6207
ChEBI CHEBI:30740
IUPAC Name 2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
SMILES C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O

Specifications

Color White
Quantity 500 g
Formula Weight 380.4
Percent Purity 0.996
Physical Form Powder
Chemical Name or Material Ethylene glycol-bis-(β-aminoethylether)-N,N,N',N'-tetraacetic acid