missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N',N'-tetraacetic Acid 96.0+%, TCI America™

Catalog No. E010625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E010625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E010625G Supplier TCI America Supplier No. E010625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 67-42-5
Molecular Formula C14H24N2O10
Molecular Weight (g/mol) 380.35
MDL Number MFCD00004291
InChI Key DEFVIWRASFVYLL-UHFFFAOYSA-N
Synonym egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
PubChem CID 6207
ChEBI CHEBI:30740
IUPAC Name 2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
SMILES C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O

Specifications

Melting Point 241°C
Color White
Quantity 25 g
Formula Weight 380.35
Percent Purity ≥96.0% (T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N′,N′-tetraacetic Acid
RTECSNumber AH3760000
TSCA Yes