missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene glycol, Analytical Standard, ≥99.9% (GC), MilliporeSigma™ Supelco™
Ethylene glycol is a colorless liquid, which may be obtained via hydration of ethylene oxide.
Supplier: Merck Emd Millipore 8597810MLF
Specifications
| 107-21-1 | |
| 2.1 (vs. air) | |
| Neat | |
| MFCD00002885 | |
| 195°C to 198°C | |
| 10 mL | |
| ≥99.9% (GC) |
| 0.08 mmHg (20°C) | |
| 1.113 g/mL (at 25°C) | |
| HOCH2CH2OH | |
| -13°C | |
| 1,2-Ethanediol | |
| 62.07 | |
| Analytical Standard |