missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, Analytical Standard, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7193405
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018710 5 mL
111018340 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111018710 Supplier Merck Emd Millipore Supplier No. 859785MLF
Only null left

Ethylene glycol is a colorless liquid, which may be obtained via hydration of ethylene oxide.

Specifications

CAS 107-21-1
Vapor Pressure 0.08 mmHg (20°C)
Vapor Density 2.1 (vs. air)
Density 1.113 g/mL (at 25°C)
Physical Form Neat
Linear Formula HOCH2CH2OH
MDL Number MFCD00002885
Melting Point -13°C
Boiling Point 195°C to 198°C
Synonym 1,2-Ethanediol
Quantity 5 mL
Formula Weight 62.07
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less