missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, 99.0%, MilliporeSigma™

Catalog No. 50947519
Change view
Click to view available options
Quantity:
3 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50947519 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50947519 Supplier Technidata Supplier No. EX05641

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.07
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
IUPAC Name ethane-1,2-diol
SMILES OCCO

Specifications

pH 6 to 7.5 (100 g/L, H2O, 20°C)
Melting Point −14 to −10°C
Quantity 3 L
Flash Point 111°C
Packaging Glass Bottle (Pre-packed)
Assay Percent Range 99.0% min
Vapor Pressure 0.053 hPa (20°C)
Boiling Point 197.6°C (1013 hPa)
Solubility Information 1000 g/L
Recommended Storage Store at room temperature