missing translation for 'onlineSavingsMsg'
Learn More

Ethylene dimethacrylate, 98%, stabilized

Catalog No. AC409922500
Change view
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC409922500 250 mL Glass bottle
AC409920010 1 L Glass bottle
AC409920051 5 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC409922500 Supplier Thermo Scientific Chemicals Supplier No. 409922500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97-90-5 | C10H14O4 | 198.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-90-5
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00008590
InChI Key STVZJERGLQHEKB-UHFFFAOYSA-N
Synonym ethylene glycol dimethacrylate, ethylene dimethacrylate, glycol dimethacrylate, diglycol dimethacrylate, ethanediol dimethacrylate, ethylenedimethyacrylate, ethylene methacrylate, ethyldiol metacrylate, 1,2-bis methacryloyloxy ethane, ethylene glycol bis methacrylate
PubChem CID 7355
ChEBI CHEBI:53436
SMILES CC(=C)C(=O)OCCOC(=O)C(C)=C

Specifications

Melting Point -40.0°C
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 98.0°C to 100.0°C (5.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4530 to 1.4550
Linear Formula (H2C=C(CH3)COOCH2-)2
Quantity 250 mL
Specific Gravity 1.05
Solubility Information Solubility in water: soluble.
IUPAC Name 2-(2-methylprop-2-enoyloxy)ethyl 2-methylprop-2-enoate
Stabilizer 90 to 110ppm MEHQ
Viscosity 3.1 mPa.s (20°C)
Formula Weight 198.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethylene dimethacrylate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
EINECSNumber 202-617-2
RTECSNumber OZ4400000
TSCA TSCA

RUO – Research Use Only