missing translation for 'onlineSavingsMsg'
Learn More

ethyle4-formyl-3,5-dimethyl-1H-pyrrole-2-carboxylate, 97%, Thermo Scientific™

Numéro de catalogue. CC23004CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC23004CB 250mg
CC23004DA 1g
CC23004DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CC23004CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC23004CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 2199-64-6
Formule moléculaire C10H13NO3
Poids moléculaire (g/mol) 195.22
Numéro MDL MFCD00030384
Clé InChI CLJUICOFPKFFGJ-UHFFFAOYSA-N
Synonyme 4-formyl-3,5-dimethyl-1h-pyrrole-2-carboxylic acid ethyl ester, ethyl 3,5-dimethyl-4-formylpyrrole-2-carboxylate, ethyl 4-formyl-3,5-dimethylpyrrole-2-carboxylate, 2,4-dimethyl-3-formyl-5-carbethoxypyrrole, 3,5-dimethyl-2-ethoxycarbonyl-4-formyl-1h-pyrrole, 3,5-dimethyl-4-formylpyrrole-2-carboxylic acid ethyl ester, ethyl4-formyl-3,5-dimethyl-1h-pyrrole-2-carboxylate, 1h-pyrrole-2-carboxylic acid, 4-formyl-3,5-dimethyl-, ethyl ester, 1h-pyrrole-2-carboxylicacid,4-formyl-3,5-dimethyl-,ethyl ester, 3,5-dimethyl-4-formyl-1h-pyrrole-2-carboxylic acid ethyl ester
PubChem CID 137486
Nom de l’IUPAC ethyl 4-formyl-3,5-dimethyl-1H-pyrrole-2-carboxylate
SOURIRES CCOC(=O)C1=C(C)C(C=O)=C(C)N1

Spécifications

Nom ou substance chimique ethyle4-formyl-3, 5-dimethyl-1H-pyrrole-2-carboxylate
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Quantité 250mg
Poids de formule 195.22
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.