missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-(2-methyl-1,3-thiazol-4-yl)acetate, 95%, Thermo Scientific™

Catalog No. p-3881328
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CD09887DA 1g
CD09887DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CD09887DA Supplier Thermo Scientific Chemicals Supplier No. CD09887DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37128-24-8
Molecular Formula C8H11NO2S
Molecular Weight (g/mol) 185.241
MDL Number MFCD00052519
InChI Key DWSMURKWFWKPBT-UHFFFAOYSA-N
Synonym ethyl 2-2-methyl-1,3-thiazol-4-yl acetate, 2-methyl-thiazol-4-yl-acetic acid ethyl ester, 2-methylthiazole-4-acetic acid ethyl ester, ethyl 2-methyl-1,3-thiazol-4-yl acetate, ethyl 2-2-methylthiazol-4-yl acetate, 4-thiazoleacetic acid,2-methyl-, ethyl ester, enamine_005349, ethyl=2-methylthiazol-4-ylacetate, 2-methylthiazol-4-yl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-methyl-, ethyl ester
PubChem CID 854815
IUPAC Name ethyl 2-(2-methyl-1,3-thiazol-4-yl)acetate
SMILES CCOC(=O)CC1=CSC(=N1)C

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 1g
Formula Weight 185.25
Percent Purity 95%
Chemical Name or Material Ethyl 2-(2-methyl-1, 3-thiazol-4-yl)acetate
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.