missing translation for 'onlineSavingsMsg'
Learn More

ethyle1-methylcyclopropane-1-carboxylate, 97%, Thermo Scientific™

Catalog No. SB01040DA
Change view
Click to view available options
Quantité:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
SB01040DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB01040DA Supplier Thermo Scientific Chemicals Supplier No. SB01040DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71441-76-4
Formule moléculaire C7H12O2
Poids moléculaire (g/mol) 128.171
Numéro MDL MFCD00001284
Clé InChI IZPYNZLFBUQGCZ-UHFFFAOYSA-N
Synonyme ethyl 1-methylcyclopropanecarboxylate, 1-methylcyclopropanecarboxylic acid ethyl ester, ethyl1-methylcyclopropanecarboxylate, cyclopropanecarboxylic acid, 1-methyl-, ethyl ester, cyclopropanecarboxylicacid, 1-methyl-, ethyl ester, ethyl 1-methylcyclopropane-carboxylate, 1-methylcyclopropanoic acid ethyl ester, 1-ethoxycarbonyl-1-methylcyclopropane, ethyl 1-methylcyclopropan-1-carboxylate, ethyl 1-methyl cyclopropane-1-carboxylate
PubChem CID 544234
Nom de l’IUPAC ethyl 1-methylcyclopropane-1-carboxylate
SOURIRES CCOC(=O)C1(CC1)C

Specifications

Nom ou substance chimique Ethyl 1-methylcyclopropane-1-carboxylate
CAS % minimum 97.0
Densité 0.9180g/mL
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Formule linéaire CH3C3H4C(O)OCH2CH3
Quantité 1g
Poids de formule 128.17
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.