missing translation for 'onlineSavingsMsg'
Learn More

Éthylcyclohexane 99,0+%, TCI America™

Numéro de catalogue. E0073500ML
Modifier l'affichage
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
E0073500ML 500 mL
E007325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. E0073500ML Fournisseur TCI America Code fournisseur E0073500ML

Identifiants chimiques

CAS 1678-91-7
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00001523
InChI Key IIEWJVIFRVWJOD-UHFFFAOYSA-N
Synonym cyclohexane, ethyl, ethyl cyclohexane, cyclohexylethane, ethyl-cyclohexane, acmc-1c1qu, ethylcyclohexane, dsstox_cid_30334, dsstox_gsid_51779, wln: l6tj a2, qspl 100
PubChem CID 15504
IUPAC Name Éthylcyclohexane
SMILES CCC1CCCCC1

Spécifications

Color Incolore
Boiling Point 132°C
Quantity 500 mL
UN Number 3295
Formula Weight 112.22
Percent Purity ≥99.0% (GC)
Physical Form Liquide
Chemical Name or Material Ethylcyclohexane
EINECSNumber (3)-2231
TSCA Yes