missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ethylboronic acid pinacol ester, 99%

Catalog No. AAH3686814
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3686814 25 g
AAH3686803 1 g
AAH3686806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3686814 Supplier Thermo Scientific™ Supplier No. H3686814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 82954-89-0 | C8H17BO2 | 156.03 g/mol

Chemical Identifiers

CAS 82954-89-0
Molecular Formula C8H17BO2
Molecular Weight (g/mol) 156.03
MDL Number MFCD19441153
InChI Key NPUBDPDASOEIOA-UHFFFAOYSA-N
Synonym 2-Ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane;
IUPAC Name 2-ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CCB1OC(C)(C)C(C)(C)O1

Specifications

Boiling Point 79-81°C / 120mbar
Quantity 25 g
Formula Weight 156.03
Percent Purity 99%
Physical Form Liquid
ShelfLife 5 Years