missing translation for 'onlineSavingsMsg'
Learn More

Ethyl viologen dibromide, 97%, Thermo Scientific™

Catalog No. AC455030010
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455030010 1g
AC455030050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455030010 Supplier Thermo Scientific Chemicals Supplier No. 455030010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 53721-12-3
Molecular Formula C14H18Br2N2
Molecular Weight (g/mol) 374.11
InChI Key LCEBDKLPALDQPV-UHFFFAOYSA-L
Synonym ethyl viologen dibromide, 1,1'-diethyl-4,4'-bipyridinium dibromide, 4,4'-bipyridinium, 1,1'-diethyl-, dibromide, ethylviologen dibromide, 1,1'-diethyl-4,4'-bipyridiniumdibromide, 1,1/'-diethyl-4,4/'-bipyridinium dibromide, 1,1-diethyl-4,4-bipyridinium dibromide, 1,1'-diethyl-4,4'-bipyridine-1,1'-diium bromide, 1-ethyl-4-1-ethyl 4-pyridyl pyridine, bromide, bromide
PubChem CID 11474262
IUPAC Name 1-ethyl-4-(1-ethylpyridin-1-ium-4-yl)pyridin-1-ium;dibromide
SMILES CC[N+]1=CC=C(C=C1)C2=CC=[N+](C=C2)CC.[Br-].[Br-]

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass Bottle
Quantity 1g
Formula Weight 374.11
Percent Purity 97%
Chemical Name or Material Ethyl viologen dibromide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only