missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Vanillin, NF, 98-101%, Spectrum™ Chemical
ET119, 121-32-4, C9H10O3
$512.42 - $32526.58
Chemical Identifiers
| CAS | 121-32-4 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.18 |
| MDL Number | MFCD00006944 |
| InChI Key | CBOQJANXLMLOSS-UHFFFAOYSA-N |
| IUPAC Name | 3-ethoxy-4-hydroxybenzaldehyde |
| SMILES | CCOC1=CC(C=O)=CC=C1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602549
|
Spectrum Chemical Mfg Cor
ET119125GM |
125 g | Amber Glass Bottle |
Each for $512.42
|
|
||||
|
18602550
|
Spectrum Chemical Mfg Cor
ET119500GM |
500 g | Amber Glass Bottle |
Each for $1,094.64
|
|
||||
|
18602551
|
Spectrum Chemical Mfg Cor
ET11925KG |
2.5 kg | Poly Pail |
Each for $3,629.06
|
|
||||
|
18602552
|
Spectrum Chemical Mfg Cor
ET11912KGBL |
12 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18602553
|
Spectrum Chemical Mfg Cor
ET11940KGBL |
40 kg | Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Ethyl Vanillin, NF is used as a flavorant. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 121-32-4 | |
| 166.18 | |
| CBOQJANXLMLOSS-UHFFFAOYSA-N | |
| CCOC1=CC(C=O)=CC=C1O |
| C9H10O3 | |
| MFCD00006944 | |
| 3-ethoxy-4-hydroxybenzaldehyde |
Specifications
| 121-32-4 | |
| 1% | |
| Amber Glass Bottle | |
| MFCD00006944 | |
| CBOQJANXLMLOSS-UHFFFAOYSA-N | |
| 3-ethoxy-4-hydroxybenzaldehyde | |
| 98 to 101% |
| 100% | |
| 76-78°C | |
| C9H10O3 | |
| 125 g | |
| CCOC1=CC(C=O)=CC=C1O | |
| 166.18 | |
| NF |