missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Vanillin, NF, 98-101%, Spectrum™ Chemical
ET119, 121-32-4, C9H10O3
Supplier: Spectrum Chemical Mfg Cor ET11912KGBL
Description
Spectrum™ Chemical Ethyl Vanillin, NF is used as a flavorant. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 121-32-4 | |
| 1% | |
| Fiber Drum | |
| MFCD00006944 | |
| CBOQJANXLMLOSS-UHFFFAOYSA-N | |
| 3-ethoxy-4-hydroxybenzaldehyde | |
| 98 to 101% |
| 100% | |
| 76-78°C | |
| C9H10O3 | |
| 12 kg | |
| CCOC1=CC(C=O)=CC=C1O | |
| 166.18 | |
| NF |
Chemical Identifiers
| 121-32-4 | |
| 166.18 | |
| CBOQJANXLMLOSS-UHFFFAOYSA-N | |
| CCOC1=CC(C=O)=CC=C1O |
| C9H10O3 | |
| MFCD00006944 | |
| 3-ethoxy-4-hydroxybenzaldehyde |