missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Trichloroacetate 97.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 167°C |
| Quantity | 500 g |
| Formula Weight | 191.43 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethyl Trichloroacetate |
Safety and Handling
| EINECSNumber | (2)-3606 |
| TSCA | Yes |