missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl trans-3-Benzoylacrylate 95.0+%, TCI America™
Supplier: TCI America B12335G
Description
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
Ethyl trans-3-Benzoylacrylate | |
Yellow | |
C12H12O3 | |
5 g | |
trans-3-Benzoylacrylic Acid Ethyl Ester | |
CCOC(=O)C=CC(=O)C1=CC=CC=C1 | |
204.225 | |
204.23 | |
Liquid |
15121-89-8 | |
185°C | |
MFCD00011533 | |
3082 | |
ACXLBHHUHSJENU-CMDGGOBGSA-N | |
ethyl (E)-4-oxo-4-phenylbut-2-enoate | |
5369605 | |
≥95.0% (GC) |
Chemical Identifiers
15121-89-8 | |
204.225 | |
ACXLBHHUHSJENU-CMDGGOBGSA-N | |
5369605 | |
CCOC(=O)C=CC(=O)C1=CC=CC=C1 |
C12H12O3 | |
MFCD00011533 | |
trans-3-Benzoylacrylic Acid Ethyl Ester | |
ethyl (E)-4-oxo-4-phenylbut-2-enoate |
Safety and Handling
EINECSNumber : (3)-4220
TSCA : No