missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trans-2-Decenoate 96.0+%, TCI America™

Catalog No. D276725ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D276725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D276725ML Supplier TCI America Supplier No. D276725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7367-88-6
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00015299
InChI Key GNJARWZWODMTDR-ZHACJKMWSA-N
Synonym ethyl trans-2-decenoate, ethyl 2-decenoate, ethyl e-2-decenoate, trans-2-decenoic acid ethyl ester, ethyl 2e-dec-2-enoate, 2-decenoic acid, ethyl ester, 2e, unii-b2k05s6l5q, 2-decenoic acid, ethyl ester, e, ethyl 2-decenoate, 2e, fema no. 3641
PubChem CID 5463904
ChEBI CHEBI:87324
IUPAC Name ethyl (E)-dec-2-enoate
SMILES CCCCCCCC=CC(=O)OCC

Specifications

Color Colorless
Boiling Point 135°C
Quantity 25 mL
Formula Weight 198.31
Percent Purity ≥96.0% (GC)
Grade Reagent
Physical Form Liquid
Chemical Name or Material Ethyl trans-2-Decenoate
EINECSNumber (2)-0798
TSCA Yes